Skip to Content
MilliporeSigma
Taurocholic Acid, Sodium Salt ULTROL® Grade, ≥98% (TLC), detergent to solubilize membrane-bound proteins in their native state

Taurocholic Acid, Sodium Salt

Taurocholic Acid, Sodium Salt ULTROL® Grade, ≥98% (TLC), detergent to solubilize membrane-bound proteins in their native state
Synonym(s):
Taurocholic Acid, Sodium Salt, Taurocholic Acid, Sodium Salt, ULTROL® Grade
Empirical Formula (Hill Notation):
C26H44NO7S · Na
CAS Number:
Molecular Weight:
537.68

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment