CA
EN
Applications
Products
Services
Support
Home
Search Results
Ca2+ increases the specific coenzyme Q10 content in Agrobacterium tumefaciens
Ca2+ increases the specific coenzyme Q10 content in Agrobacterium tumefaciens
Bioprocess and Biosystems Engineering (2009)
Ha SJ, et al.