  • Search Results
  • Development of a porcine renal extracellular matrix scaffold as a platform for kidney regeneration.
Development of a porcine renal extracellular matrix scaffold as a platform for kidney regeneration.
Journal of biomedical materials research. Part A (2014-07-22)
Seock Hwan Choi, So Young Chun, Seon Yeong Chae, Jin Rae Kim, Se Heang Oh, Sung Kwang Chung, Jin Ho Lee, Phil Hyun Song, Gyu-Seog Choi, Tae-Hwan Kim, Tae Gyun Kwon
ABSTRACT
Acellular scaffolds, possessing an intact three-dimensional extracellular matrix (ECM) architecture and biochemical components, are promising for regeneration of complex organs, such as the kidney. We have successfully developed a porcine renal acellular scaffold and analyzed its physical/biochemical characteristics, biocompatibility, and kidney reconstructive potential. Segmented porcine kidney cortexes were treated with either 1% (v/v) Triton X-100 (Triton) or sodium dodecyl sulfate (SDS). Scanning electron microscopy showed both treatments preserved native tissue architecture, including porosity and composition. Swelling behavior was higher in the Triton-treated compared with the SDS-treated scaffold. Maximum compressive strength was lower in the Triton-treated compared with the SDS-treated scaffold. Attenuated total reflective-infrared spectroscopy showed the presence of amide II (-NH) in both scaffolds. Furthermore, richer ECM protein and growth factor contents were observed in the Triton-treated compared with SDS-treated scaffold. Primary human kidney cell adherence, viability, and proliferation were enhanced on the Triton-treated scaffold compared with SDS-treated scaffold. Following murine in vivo implantation, tumorigenecity was absent for both scaffolds after 8 weeks and in the Triton-treated scaffold only, glomeruli-like structure formation and neovascularity were observed. We identified 1% Triton X-100 as a more suitable decellularizing agent for porcine renal ECM scaffolds prior to kidney regeneration.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MOPS, ≥99.5% (titration)
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
SAFC
BIS-TRIS
SAFC
MOPS
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)