The kampo medicine Daikenchuto inhibits peritoneal fibrosis in mice.
Biological & pharmaceutical bulletin (2015-03-10)
Mineaki Kitamura, Tomoya Nishino, Yoko Obata, Satoru Oka, Shinichi Abe, Kumiko Muta, Yoshiyuki Ozono, Takehiko Koji, Shigeru Kohno
ABSTRACT
Long-term peritoneal dialysis therapy causes inflammation and histological changes in the peritoneal membrane. Inflammation generally activates fibroblasts and results in fibroblast-myofibroblast differentiation. Heat-shock protein 47 (HSP 47), a collagen-specific molecular chaperone, is localized in myofibroblasts and is involved in the progression of peritoneal fibrosis. Daikenchuto (DKT), a Kampo medicine, is used to prevent postoperative colon adhesion. It inhibits inflammation and HSP 47 expression in the gastrointestinal tract. We examined the effect of DKT on chlorhexidine gluconate (CG)-induced peritoneal fibrosis in mice injected with 0.1% CG dissolved in 15% ethanol. DKT was dissolved in the drinking water. Histological changes were assessed using Masson trichrome staining. Cells expressing α-smooth muscle actin (α-SMA), HSP 47, phospho-Smad 2/3, F4/80, and monocyte chemotactic protein-1 were examined immunohistochemically. Compared with the control group, the peritoneal tissues of the CG group were markedly thickened, and the number of cells expressing α-SMA, HSP 47, phospho-Smad 2/3, F4/80, and monocyte chemotactic protein-1 was significantly increased. However, these changes were inhibited in the DKT-treated group. These results indicate that DKT can prevent peritoneal fibrosis by inhibiting inflammation and HSP 47 expression.
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
SAFC
MOPS
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)