  • Search Results
  • Myocardial infarction accelerates glomerular injury and microalbuminuria in diabetic rats via local hemodynamics and immunity.
Myocardial infarction accelerates glomerular injury and microalbuminuria in diabetic rats via local hemodynamics and immunity.
International journal of cardiology (2014-12-03)
Zhifeng Dong, Kaizheng Gong, Dong Huang, Wei Zhu, Wanfeng Sun, Ying Zhang, Ping Xin, Yuan Shen, Penglong Wu, Jingbo Li, Zhigang Lu, Xiaoming Zhang, Meng Wei
ABSTRACT
Clinically, approximately one-third of patients with chronic heart failure (CHF) exhibit some degree of renal dysfunction. This renal dysfunction is referred to as cardiorenal syndrome (CRS) and plays an important role in the poor prognosis of CHF. Mounting evidence suggests that diabetes is the most common underlying risk factor for CRS. However, the underlying pathophysiological mechanisms are poorly understood. We performed the following comparisons in two separate protocols: 1) surgically induced myocardial infarction rats (MI, n=10), sham operation rats (Ctr, n=10) and MI rats treated with Fasudil, a Rho-kinase inhibitor (MI+Fas, n=9); and 2) STZ-induced type 1 diabetic rats (DB, n=10), DB+MI rats (n=10) and DB+MI rats treated with Fasudil (DB+MI+Fas, n=9). Renal hemodynamics and vasoconstrictor reactivity were evaluated using the DMT myograph system. Renal immunity was evaluated by flow cytometry, electron microscopy, immunofluorescence, etc. Twelve weeks after the operation, compared with DB or MI rats, DB+MI rats exhibited the following characteristics: 1) significantly increased glomerular enlargement, fibrosis, glomerulosclerosis, podocyte injury and microalbuminuria; 2) significantly increased vasoconstrictor reactivity of the renal interlobular arteries and renal venous pressure; 3) significantly increased infiltration of CD₃+ and CD₄+ T cells and decreased Treg/Th17 ratios; and 4) significantly increased glomerular deposition of IgG and C₄. In contrast, rats with MI only showed mildly accelerated glomerular remodeling and microalbuminuria, with little change in renal hemodynamics and immunity. Fasudil treatment significantly improved the renal lesions in DB+MI rats but not MI rats. Post-MI cardiac dysfunction significantly accelerated glomerular remodeling, podocyte injury and microalbuminuria in STZ-induced diabetic rats. These changes were accompanied by altered local hemodynamics and immunity.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture