  • Search Results
  • Ovarian microcystic stromal tumor: A novel extracolonic tumor in familial adenomatous polyposis.
Ovarian microcystic stromal tumor: A novel extracolonic tumor in familial adenomatous polyposis.
Genes, chromosomes & cancer (2015-03-31)
Sang Hun Lee, Young Wha Koh, Hyun Jin Roh, Hee Jeong Cha, Yong-Soon Kwon
ABSTRACT
Ovarian microcystic stromal tumor (MCST) is a very rare neoplasm; hence, its nomenclature was recently designated as "Distinctive morphologic and immunohistochemical features" in 2009. Its exact origin, etiological genetic alterations, and background are not yet clearly known. Familial adenomatous polyposis (FAP) is an autosomal dominant disease that leads to development of colorectal polyps via germ-line mutations of the APC gene on chromosome 5q21∼22. In this study, we report a 40-year-old female patient who had ovarian MCST and FAP. On sequencing the APC gene in ovarian MSCT, we detected a novel somatic mutation of the APC gene in exon 11, with a heterozygous deletion at nucleotide position c.1540delG (p.Ala514 Profs*9). Mutations of β-catenin (CTNNB1) and FOXL2 were not detected. Although one case demonstrating involvement of Wnt/β-catenin in ovarian MCST associated with FAP has been presented previously, no detailed information was provided. Thus, this is the ovarian MCST with a somatic mutation of APC in a patient with FAP.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)