  • Search Results
  • Feasibility of antibody-poly(glutamic acid) complexes: preparation of high-concentration antibody formulations and their pharmaceutical properties.
Feasibility of antibody-poly(glutamic acid) complexes: preparation of high-concentration antibody formulations and their pharmaceutical properties.
Journal of pharmaceutical sciences (2015-03-27)
Shunsuke Izaki, Takaaki Kurinomaru, Takuya Maruyama, Takayuki Uchida, Kenji Handa, Tomoaki Kimoto, Kentaro Shiraki
ABSTRACT
Development of high-concentration antibody formulations for subcutaneous administration remains challenging. Recently, a precipitation-redissolution method was proposed to prepare suspensions or precipitates of salt-dissociable protein-poly(amino acid) complexes. To elucidate the utility of this method for protein therapy, we investigated the feasibility of a precipitation-redissolution method using poly(amino acid) for high-concentration antibody formulation. Omalizumab and adalimumab formulations of 150 mg/mL could be prepared using poly-l-glutamic acid (polyE) from low-concentration stock solutions. Enzyme-linked immunosorbent assay, circular dichroism, and size-exclusion chromatography revealed that the formation of antibody-polyE complex and precipitation-redissolution process did not significantly affect the immunoreactivity or secondary structure of the antibodies. The precipitation-redissolution method was less time-consuming and more effective than lyophilization-redissolution, evaporation-redissolution, and ultrafiltration from the viewpoint of final yield. Scalability was confirmed from 400 μL to 1.0 L. The general toxicity and pharmacokinetic profiles of the antibody-polyE complex formulations were similar to those of conventional antibody formulations. These results suggested that the precipitation-redissolution method using poly(amino acid) has great potential as a concentration method for antibody formulation and medicinal use.
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis