  • Search Results
  • Supplementary studies and the first molecular data on Myxobolus scardinii Reuss, 1906 (Myxozoa: Myxosporea) infecting the gill filaments of rudd, Scardinius erythrophthalmus (L.).
Supplementary studies and the first molecular data on Myxobolus scardinii Reuss, 1906 (Myxozoa: Myxosporea) infecting the gill filaments of rudd, Scardinius erythrophthalmus (L.).
Parasitology research (2015-06-22)
Gokmen Zafer Pekmezci, Banu Yardimci, Savas Yilmaz
ABSTRACT
Within the present study, we offer new information on the morphology, histopathology, and 18S ribosomal DNA (rDNA) sequence of Myxobolus scardinii to supplement and revise the original description of Reuss (Bull Acad Imp Sci St-Pétersbourg, V Serie, XXV, No. 3: 199-205, 1906). M. scardinii was found infecting the gills of 6 of 125 specimens (4.8%) of Scardinius erythrophthalmus from northern Turkey. Large mature plasmodia, whitish, spherical, or ellipsoidal, measuring 0.9-1.0 mm in diameter, macroscopically occurred in the gills. Histopathological examination indicated that development of the cyst-like plasmodia was intrafilamental-vascular type. Mature spores are oval or shortly ellipsoidal. The spores are 12.1 ± 0.2 (11.5-12.6) μm long, 8.9 ± 0.3 (8.4-9.2) μm wide, and 6.1 ± 0.3 (5.9-6.2) μm thick. The two polar capsules are equal in size, 4.8 ± 0.2 (4.7-5.1) μm long and 2.8 ± 0.2 (2.7-3.1) μm wide, and are long pyriform in shape. A well-developed intercapsular appendix was found in the fresh spores. In this study, a specific gill myxosporean of S. erythrophthalmus, M. scardinii the identity of which was confirmed by careful morphological and histopathological examination, was sequenced with its 18S rDNA sequence for the first time and submitted to Genbank database (accession number KJ562362). Phylogenetic analysis of the 18S rDNA gene revealed that M. scardinii had the closest similarity to M. muelleri and M. bramae. This is the first molecular data for the validity of M. scardinii in S. erythrophthalmus. This new valid genetic data (KJ562362) has been used to establish phylogenetic relationships with other similar gills Myxobolus species from Eurasia.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, 98%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M