  • Search Results
  • FIZZ1 Promotes Airway Remodeling in Asthma Through the PTEN Signaling Pathway.
FIZZ1 Promotes Airway Remodeling in Asthma Through the PTEN Signaling Pathway.
Inflammation (2015-02-07)
Jiping Zhao, Xingai Jiao, Jinxiang Wu, Junfei Wang, Wenbin Gong, Fen Liu, Wen Liu, Wenxiang Bi, Liang Dong
ABSTRACT
The aim of our study was to elucidate the function and signaling pathway of found in inflammatory zone 1 (FIZZ1) in airway remodeling in asthma. We used a mice model sensitized and challenged by ovalbumin (OVA) to evaluate the expression of FIZZ1, type I collagen, and fibronectin-1 in the airway in asthma. To investigate the signaling pathway regulated by FIZZ1, we treated a cultured murine lung epithelium cell-12 (MLE-12) with FIZZ1 recombination protein, silenced the expression of FIZZ1 with FIZZ1-shRNA in vitro, and then detected phosphorylated phosphatase and tensin homolog deleted on chromosome 10 (PTEN) and expression of type I collagen and fibronectin-1 (FN-1) by Western blotting. In addition, we increased the expression of PTEN by PTEN plasmid transfection then detected the expression of type I collagen and fibronectin-1 in MLE-12 by Western blot analysis and immunofluorescence cytochemistry technology, respectively. First, the expression of FIZZ1, type I collagen, and fibronectin-1 was significantly elevated in the lungs of OVA-challenged mice compared with saline-treated control animals. Secondly, the phosphorylation of PTEN was decreased in MLE-12 treated with FIZZ1 recombination protein in vitro. On the contrary, the phosphorylation of PTEN was increased in MLE-12 cells transfected with FIZZ1-shRNA. Thirdly, results of the Western blot analysis and immunofluorescence cytochemistry showed that expression of type I collagen and fibronectin-1 was increased in cells treated with FIZZ1 recombination protein, while the levels of type I collagen and fibronectin-1 were significantly decreased in cells transfected with PTEN plasmid. FIZZ1 may be a critical cytokine in airway remodeling in asthma. This study indicates that targeting FIZZ1 and/or PTEN may be a new therapeutic strategy for asthma.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
SAFC
BIS-TRIS
SAFC
MOPS
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl