  • Search Results
  • Detection of fraudulent addition of bovine whey in water buffalo ricotta cheese by isoelectric focusing.
Detection of fraudulent addition of bovine whey in water buffalo ricotta cheese by isoelectric focusing.
Journal of the science of food and agriculture (2014-11-27)
Fabio Fuselli, Anna Deluca, Emanuela A Montepeloso, Giulia Ibba, Flavio Tidona, Lucia Longo, Rosa M Marianella
ABSTRACT
Prevention of food fraud in the dairy field is a difficult issue for researchers, industries and policy makers, both for commercial and health reasons. Currently, no analytical method allows detection of the addition of bovine whey to water buffalo ricotta, so this fraudulent practice cannot be prevented. The authors' aim was to develop such a method. The conditions for extraction and purification of denatured ricotta whey proteins, which are unfolded and coagulated by heating during the production process, were optimized. The optimal composition of the polyacrylamide gel (pH range, type and concentration of chemical separator) was first evaluated and then the best conditions to perform the separation by isoelectric focusing were established. The performance of the method (precision, selectivity, robustness, sensibility) was determined. The method was shown to be reliable and robust for detection of the presence of bovine whey added to water buffalo Ricotta at percentages above 5% (v/v). The results suggest that the differences observed between bovine and water buffalo electrophoretic profiles are due to bovine β-lactoglobulin isoform A, which is never detected in water buffalo samples.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ~99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%