  • Search Results
  • PAMAM G4 dendrimers as inhibitors of the iron storage properties of human L-chain ferritin.
PAMAM G4 dendrimers as inhibitors of the iron storage properties of human L-chain ferritin.
Physical chemistry chemical physics : PCCP (2015-07-02)
M B Camarada, V Márquez-Miranda, I Araya-Durán, A Yévenes, F González-Nilo
ABSTRACT
Cationic dendrimers, such as PAMAM, are known to be positively charged at neutral pH allowing their unspecific interaction with proteins and other cellular components. Especially, ferritin, which has an important role in iron homeostasis, presents a negative electrostatic potential at the 3-fold channel. This channel is important in the functionality of ferritin because it allows the iron entry into its inner cavity. In this way, the interaction between the protonated terminal amines of the dendrimer and the negatively charged 3-fold channels of ferritin is expected. Experimental measurements demonstrated that PAMAM G4 inhibits the iron storage properties of L-chain human ferritin (L-Ftn). Molecular dynamics simulations have been used to analyze the specific interaction between PAMAM G4 and L-Ftn. Results show that PAMAM G4 effectively interacts with the 3-fold channels of L-Ftn, suggesting that this interaction is responsible for the inhibition of the iron storage properties of L-Ftn.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Thioglycolic acid, 98%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)