  • Search Results
  • Regulatory role of charged clusters in the N-terminal domain of BetP from Corynebacterium glutamicum.
Regulatory role of charged clusters in the N-terminal domain of BetP from Corynebacterium glutamicum.
Biological chemistry (2015-07-07)
Izabela Waclawska, Christine Ziegler
ABSTRACT
The trimeric transporter BetP counteracts hyperosmotic stress by a fast increase in transport rate in order to accumulate the compatible solute betaine. The positively charged α-helical C-terminal domain acts as an osmosensor perceiving the increase in the internal potassium (K+) concentration. A second, still unidentified stimulus originates from stress-induced changes in the physical state of the membrane and depends on the amount of negatively charged lipids. BetP possesses a 60-amino acid (aa)-long negatively charged N-terminal domain, which is predicted to adopt a partly helical fold affecting osmoregulation by an unknown mechanism. It is assumed that the C-terminal domain, the N-terminal domain, and negatively charged lipids interact during stress sensing and regulation. Here, we have investigated the regulatory role of negatively charged clusters in the N-terminal domain. We identified one cluster, Glu24Glu25, to be crucial for osmoregulation. Cross-linking studies revealed an interaction between the C- and N-terminal domains of adjacent protomers modulating transport activation. A regulatory partner-switching mechanism emerges in which the C-terminal domain changes its interaction with the N-terminal domain of its own promoter and negatively charged lipids to an interaction with the N-terminal domain of an adjacent protomer and lipids bound to the central cavity of the BetP trimer.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride solution, 0.5 M in anhydrous tetrahydrofuran
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Potassium chloride, meets EP, USP testing specifications
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis