MilliporeSigma
HomeProduct SupportingBiochemicalsFluorescent Lipid Probes - Avanti Research™
Fluorescent Lipid Probes - Avanti Research™
Illuminate Your Research with Fluorophores
Polar bear swimming underwater with white fur contrasting blue water, surrounded by bubbles and light reflecting off the surface above.
Figure 1.Discover Avanti Research™ fluorophores.
Fluorescent probes are essential tools for visualizing biological systems, enabling researchers to track molecules, monitor disease markers, and study cellular function with high sensitivity and minimal disruption. By absorbing and emitting light at specific wavelengths, these fluorophores reveal valuable insights, even at low concentrations.
As the exclusive partner for Avanti Research™ products outside the U.S. and Canada, we offer one of the most comprehensive selections of fluorescent and lipid-conjugated probes, spanning the full emission spectrum. The portfolio includes widely used fluorophores such as FITC, Rhodamine, TopFluor™ (BODIPY), NBD, Cy3, Cy5, and more, giving you the flexibility to target a broad range of biomolecules across diverse applications.
Explore our fluorescent probe solutions and bring your research into focus. With optimized fluorescence, achieve bright signals and clear, confident results.
Three colorful molecular models of fluorescent lipids: linear (a), branched (b), and stacked (c), with color-coded atoms and functional groups.
Figure 2.Three examples of offerings in our ever growing fluorescent lipids portfolio: a) TopFluor™ Cholesterol; b) TopFluor™ PE; and c) TopFluor™ Cardiolipin
Fluorescent Probes:
Nitrobenzoxadiazole (NBD)
Environmentally sensitive and easily modified, NBD reacts with amines and thiols, making it ideal for labeling lipids, studying membrane dynamics, and probing biological nucleophiles in live systems.
Black-on-white structure of NBD shows a benzene ring with diazole, nitro, and amine groups; R group in blue indicates a variable substituent.
Figure 3.Structure of fluorophore nitrobenzoxadiazole (NBD).
TopFluor™ (BODIPY)
TopFluor™ (BODIPY) is a highly versatile, photostable fluorophore with tunable emission (500–700 nm) and strong quantum yields across environments. Its minimal impact on lipid behavior makes TopFluor™-labeled lipids ideal for studying membrane dynamics, lipid diffusion, and cellular localization.
Black-on-white  structure of Top Fluor™ shows  a fluorinated aromatic ring with  F-labeled substituents, highlighting  its fluorescent molecular design.
Figure 4.Structure of Top Fluor™.
Black-on-white structure of Top Fluor TMR™ shows a boron-fluorine complex with aromatic rings, a carbonyl group, and a blue R group for variability.
Figure 5.Structure of Top Fluor TMR™.
Cyanines
Cyanine probes are fluorescent dyes composed of two indole rings linked by a variable-length polyalkene chain, with longer chains shifting excitation and emission to longer wavelengths. Known for their high photostability and water solubility, cyanine dyes are ideal for labeling amines on proteins, antibodies, and nucleic acids for detection via microscopy or flow cytometry.
Black-on-white structure of Cyanine n.5 shows two aromatic rings linked by a variable carbon chain, with charged N, Cl⁻, and a blue R group for variability.
Figure 6.Structure of fluorescent probe cyanine n.5, where n = carbon linker length.
Black-on-white structure of Cyanine n shows aromatic rings linked by a variable carbon chain, with charged N, Cl⁻, and a blue R group for substitution.
Figure 7.Structure of fluorescent probe cyanine n, where n = carbon linker length.
SquareFluor™ (Squaraines)
SquareFluor™ (squaraine) probes feature a squaric acid-derived aromatic ring that enables intense absorption, strong photostability, and high quantum yield. Once limited by sensitivity to nucleophilic attack, recent design improvements have renewed interest in their use for bioimaging, photodynamic therapy, and analyte sensing.
Structure features a square fluorophore with aromatic rings, charged nitrogen, and a blue R group indicating variable substitution.
Figure 8.Structure of fluorescent probe SquareFluor™ (Ex/Em: 610/630 nm).
Lissamine Rhodamine (Liss Rhod)
Lissamine Rhodamine (Liss Rhod) is a fluorescent probe commonly used to study cell membrane dynamics, membrane fusion, and liposome behavior. Its strong fluorescence makes it ideal for visualizing lipid interactions, with tagged lipids like PEs, fatty acids, and triglycerides available for a range of applications.
Black-on-white structure shows fused aromatic rings, charged nitrogen, a sulfonate group, and a blue R group for variable substitution.
Figure 9.Structure of fluorescent probe Lissamine Rhodamine (Liss Rhod).
DPH
1,6-Diphenyl-1,3,5-hexatriene (DPH) is a hydrophobic fluorescent probe widely used to study membrane dynamics and lipid bilayer structure via fluorescence anisotropy. Its weak fluorescence in water but strong emission in hydrophobic environments makes it ideal for assessing lipid tail order, membrane fluidity, and interfacial properties.
Structure shows a linear hexatriene chain with phenyl rings at each end, a carbonyl group, and a blue R group for variable substitution.
Figure 10.Structure of fluorescent probe DPH (1,6-Diphenyl-1,3,5-hexatriene).
Pyrene
Pyrenedecanoyl (pyrene) is a widely used fluorescent label known for its ability to form excimers, excited-state dimers with red-shifted emission around 470 nm. This unique property makes pyrene-labeled lipids ideal for studying membrane fusion, phospholipid transfer, protein folding, and structural changes in biomolecules.
Structure shows a four-ring fused aromatic system with a blue R group attached, indicating a variable substituent on the pyrene core.
Figure 11.Structure of fluorophore pyrenedecanoyl (pyrene).
Dansyl
Dansyl probes are fluorescent labels often used to modify proteins and amino acids, offering high quantum yield and large Stokes shifts for easy detection. They are also useful in heavy metal sensing due to electron transfer interactions with metal ions.
Structure shows fused aromatic rings with a sulfonyl group, alkyl-linked nitrogen, and a blue R group for variable substitution.
Figure 12.Structure of fluorophore danysl.
Laurdan
Laurdan is a polarity-sensitive fluorescent probe whose emission spectrum shifts based on the polarity of its surrounding environment. This unique responsiveness makes it a powerful tool for investigating membrane order and lipid bilayer dynamics.
Structure shows a long hydrocarbon chain linked to a carbonyl and two-ring aromatic system ending in a nitrogen substituent.
Figure 13.Structure of fluorescent probe laurdan.
Fluorescein
Fluorescein is a widely used fluorescent probe known for its strong green emission and versatility in biological applications. Beyond its use in ophthalmology, it aids in bioimaging and tissue differentiation, with derivatives like FITC commonly used for protein labeling and antibodies.
Black line drawing of fluorescein showing key groups: carbonyl, ammonium, hydroxyl, and variable "R" on two aromatic rings.
Figure 14.Structure of fluorescent probe fluorescein.
Azide functionalized fluorescent probes
Azide-functionalized fluorescent probes enable copper-catalyzed click chemistry reactions with other molecules containing “acceptor” groups, such as alkynes.
Black line drawing of azide-functionalized molecule showing linear chain and azide group with N₃ atoms bearing + and − charges.
Figure 15.Structure of an azide-functionalized molecule.
Maleimide functionalized fluorescent probes
Maleimide is especially effective for conjugating fluorescent probes to proteins or peptides with cysteine residues, offering a selective and efficient method to attach the probe to molecules containing thiol functional groups.
Black line drawing of maleimide showing a five-membered ring with nitrogen, two opposite carbonyls, and a wavy line above the nitrogen.
Figure 16.Structure of fluorophore maleimide.
DBCO functionalized fluorescent probes
Fluorescent probes functionalized with DBCO enable copper-free click chemistry by reacting with molecules that have “donor” groups like azides.
Black line drawing of DBCO showing three fused aromatic rings with a central nitrogen and a wavy line beneath it indicating a possible bond.
Figure 17.Structure of DBCO.
NHS functionalized fluorescent probes
Featuring an activated NHS ester moiety, these fluorophores readily react with nucleophiles like amines. NHS esters can bind to amines under mild alkaline conditions without needing additional activation. These probes are commonly used for fluorescent labeling of peptides or proteins.
Black line drawing of NHS ester showing a five-membered ring with nitrogen, two carbonyls, and an ester group with a wavy line above.
Figure 18.Structure of an NHS ester moiety.
Free acid functionalized fluorescent probes
Fluorescent probes functionalized with free acid groups contain a carboxylic acid moiety that reacts with primary amines to form stable amide bonds.
Black line drawing of a free acid group showing a carbonyl and hydroxyl attached to a carbon, with a wavy line indicating a possible bond.
Figure 19.Structure of a free acid group.
Vertical color gradient from violet to red showing emission peaks of fluorescent compounds like pyrene, DPH, and laurdan with wavelength labels.
Figure 20.Emission Spectrum Guide provided by Avanti Research™.
Discover the complete fluorescent lipid portfolio here.
Explore our full range of biochemical solutions.
*All chemical structures provided by Avanti Research™.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500