MilliporeSigma
HomeResearch & Disease AreasCarbohydrates as Versatile Enablers of Advanced Drug Delivery Systems
Carbohydrates as Versatile Enablers of Advanced Drug Delivery Systems
Dr. Sai Sandeep Mannimala, Global Product Manager, MilliporeSigma
Carbohydrates have advanced from simple excipients to essential materials in modern drug delivery systems. Their structural diversity, gel-forming capacity, and interactions with biological interfaces enable precise control of drug release, stability, and targeting. Polysaccharides such as cellulose derivatives, chitosan, alginate, pectin, and dextran support a wide range of strategies, including matrix-controlled release, nanocarriers, mucoadhesive platforms, permeation enhancers, and site-specific systems.
Recent progress in stimuli-responsive polymers, protein and nucleic acid stabilization, and injectable depots further underscores their role in addressing complex formulation challenges. This review highlights established carbohydrate mechanisms and emerging opportunities in biologics, mRNA therapeutics, and personalized medicine, positioning carbohydrates as central to next-generation pharmaceutical technologies.
Explore our complete portfolio of carbohydrate research materials, curated to advance formulation studies. All products referenced are intended for research use only. To gain deeper insights and application information, please fill out our web form.
Close-up of a gloved hand holding a transparent red pill, with a blurred woman’s face examining it in the background indicating a clinical or scientific setting.
List of Carbohydrates for Your Research
Explore our comprehensive selection of carbohydrates, including cellulose, starch, chitosan, and marine polysaccharides, tailored for your research needs. 
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Proven Carbohydrate Drug Delivery Mechanisms
Carbohydrates play a crucial role in enhancing drug delivery systems through various innovative mechanisms. By leveraging their unique properties, these systems enable precise control over drug release, stability, and targeting.
Matrix-Controlled Release Systems
Modern carbohydrate matrices create dynamic gel barriers through polymer hydration and swelling. For instance, HPMC tablets form gel layers that control drug diffusion, with molecular weight distribution critically influencing release profiles. Recent research reveals that broader molecular weight distributions create more robust gel layers with improved consistency.1 Mathematical modeling has evolved from simple diffusion equations to sophisticated multi-phase models that account for gel layer dynamics, enabling formulators to predict release profiles and optimize formulations with greater precision.2
Carbohydrate Nanocarriers and Nanotechnology-Based Delivery
Carbohydrate-coated nanoparticles represent one of the most rapidly advancing areas, with dextran-coated iron oxide nanoparticles achieving FDA approval.3 Chitosan nanoparticles demonstrate enhanced cellular uptake through electrostatic interactions with negatively charged cell membranes, while alginate nanoparticles provide pH-responsive release behavior. Self-assembled nanostructures from amphiphilic carbohydrate derivatives create stable drug carriers with controlled release properties.4
Polymeric nanoparticles from natural carbohydrates offer biodegradable alternatives to synthetic systems. Chitosan-based nanoparticles show particular promise for gene delivery applications, while pectin nanoparticles enable colon-specific targeting through bacterial enzyme degradation.5
Electrospun nanofiber systems offer high surface area matrices that enhance drug loading capacity and controlled release. Nanofibers composed of cellulose and chitosan have demonstrated improved transdermal drug permeation, with fiber diameter playing a critical role in delivery efficiency.6
Stimuli-Responsive and Smart Delivery Systems
Multi-stimuli responsive platforms integrate multiple triggering mechanisms for enhanced specificity. pH-responsive chitosan systems swell in acidic environments while remaining collapsed at neutral pH, enabling gastric-specific delivery.
Temperature-responsive systems utilize phase transition temperatures of hydroxypropyl cellulose (37°C) for injectable gels that solidify at body temperature.8
Enzyme-responsive systems incorporate enzymatically cleavable linkages that respond to specific enzymes at target sites. Pectin-based systems utilize pectinase enzymes produced by colonic bacteria, while chitosan systems respond to lysozyme in inflammatory conditions.9
Magnetic field-responsive systems combine carbohydrate matrices with magnetic nanoparticles for externally controlled drug release. Redox-responsive delivery exploits the reducing environment inside cells, with disulfide-linked carbohydrate conjugates releasing drugs upon cellular uptake.10
Mucoadhesive and Bioadhesive Mechanisms
Mucoadhesive systems exploit the ability of cationic carbohydrates like chitosan to interact electrostatically with negatively charged mucin. The strength and duration of mucoadhesion depend on polymer molecular weight, charge density, and specific mucus composition at the target site. Thiolated chitosans form disulfide bonds with mucus proteins, significantly extending residence time.11
Bioadhesive patches utilize carbohydrate-based adhesives that maintain intimate contact with biological surfaces while controlling drug release. Recent innovations include pressure-sensitive adhesives based on modified carbohydrates that provide both adhesion and controlled release functionality.12
Permeation Enhancement Mechanisms
Tight junction modulation represents a key mechanism by which chitosan enhances drug permeation across epithelial barriers. Chitosan interacts with tight junction proteins, causing reversible opening that allows paracellular transport of hydrophilic drugs. The effect is concentration and molecular weight dependent, with optimal enhancement occurring at specific chitosan concentrations.13
Improves transcellular permeation by interacting with cell membrane phospholipids. Chitosan temporarily increases membrane fluidity, facilitating transcellular drug transport across epithelial barriers.14 Follicular targeting exploits hair follicles as drug reservoirs, with carbohydrate nanoparticles showing preferential accumulation in follicular openings.15
Site-Specific and Colon-Targeted Delivery
Microbiome-triggered release exploits the unique enzymatic environment of the colon. Resistant starch and pectin systems remain intact in the upper GI tract but are rapidly degraded by colonic bacteria, providing site-specific drug release. Enzyme-specific degradation enables targeting diseased tissues with elevated enzyme activity.16
pH-gradient responsive systems utilize the natural pH variation throughout the GI tract. Enteric-coated systems using carbohydrate-based polymers protect drugs from gastric acid while enabling release in the alkaline environment of the small intestine.16
Protein and Biologics Delivery
The delivery of proteins and biologics is critical for effective therapeutic applications, and carbohydrates play a key role in their stability and efficacy.
Protein stabilization mechanisms involve carbohydrates forming protective hydrogen bonds during dehydration processes. Trehalose and sucrose create glassy matrices that maintain protein structure during freeze-drying and storage.17 Vaccine delivery systems utilize carbohydrate targeting to enhance antigen uptake by antigen-presenting cells, improving immune responses.18
Gene delivery platforms based on cationic carbohydrates like chitosan and DEAE-dextran can condense DNA/RNA through electrostatic interactions, protecting nucleic acids from degradation while enhancing cellular uptake.19,20 Antibody conjugation to carbohydrate carriers improves antibody stability and enables controlled release.21
Injectable and Implantable Systems
Injectable and implantable carbohydrate-based systems provide advanced solutions for sustained drug delivery, enabling controlled release that adapts to physiological conditions.
In-situ gel formation occurs when carbohydrate solutions undergo gelation upon injection due to physiological conditions. Alginate systems gel in the presence of calcium ions, while chitosan systems respond to physiological pH changes. These systems provide sustained drug release over weeks to months8.
Microsphere encapsulation using carbohydrate polymers enables controlled drug release through diffusion and polymer degradation. Alginate microspheres created through ionic crosslinking provide tunable release rates based on polymer concentration and crosslinking density.22
Implantable carbohydrate matrices utilize carbohydrate matrices for long-term drug delivery. Biodegradable implants based on modified starches and cellulose derivatives provide sustained release over months while gradually degrading to non-toxic products.23
Transdermal and Topical Enhancement
Carbohydrates enhance transdermal and topical drug delivery by improving skin absorption through techniques such as occlusion, iontophoresis, and microneedle systems.
Skin penetration enhancement mechanisms include occlusive effects that increase skin hydration, direct interaction with skin lipids, and follicular targeting.13,15 Carbohydrate-based patches create optimal conditions for drug permeation while maintaining skin integrity.
Iontophoretic enhancement utilizes the ionic nature of certain carbohydrates to facilitate electrically assisted drug delivery.24 Microneedle systems coated with carbohydrate matrices enable painless drug delivery through the skin barrier, with rapid dissolution providing immediate drug release.25
Solubility Enhancement and Complexation
Carbohydrates such as cyclodextrins and micellar systems play a key role in enhancing solubility and bioavailability in drug formulations.
Cyclodextrin inclusion complexes achieve multi-fold solubility enhancement through molecular encapsulation. Ternary systems combining cyclodextrins with water-soluble polymers create synergistic solubilization effects that exceed simple inclusion complexes.26
Micellar systems from amphiphilic carbohydrate derivatives create drug-loaded micelles with enhanced bioavailability and controlled release properties.27
Emerging Applications and Research Opportunities for Carbohydrates in Pharmaceuticals
Emerging research is uncovering new roles for carbohydrates in advancing pharmaceutical science. Beyond their established uses, carbohydrate-based systems are now being explored in AI-driven formulation design, biologics and mRNA stabilization, and personalized medicine. These innovations highlight the versatility of carbohydrate materials in enabling faster development, targeted delivery, and patient-specific therapies.
AI-Driven Formulation Design
Machine learning algorithms trained on carbohydrate structure-property relationships can predict material behavior and optimize formulations with minimal experimental work. Early applications show a significant reduction in development timelines with improved success rates.28
Biologics and mRNA Applications
The expanding biologics market creates unprecedented opportunities for carbohydrate stabilization systems. mRNA therapeutics represent emerging applications where carbohydrate systems may play crucial roles in targeted delivery and protection from degradation.29,30
Personalized Medicine Integration
Carbohydrate systems can be tailored for individual patient characteristics, disease states, and physiological conditions. Manufacturing technologies like 3D printing and microfluidic patterning now allow the fabrication of personalized oral tablets, transdermal patches, and implants using materials like HPMC, ethyl cellulose, maltitol, maltodextrin, and sodium alginate.31
Conclusion
Carbohydrates have evolved from simple excipients to integral components of advanced drug delivery systems. Their structural diversity, biocompatibility, and capacity for targeted delivery enable solutions to complex formulation challenges, including biologics, mRNA therapeutics, and personalized medicine. As formulation strategies continue to advance, careful selection and application of carbohydrate systems will remain critical for achieving reproducible, effective, and tailored therapeutic outcomes.
Explore our full portfolio of carbohydrate products to support advanced formulation studies.
References
1.
Jain AK, Söderlind E, Viridén A, Schug B, Abrahamsson B, Knopke C, Tajarobi F, Blume H, Anschütz M, Welinder A, et al. 2014. The influence of hydroxypropyl methylcellulose (HPMC) molecular weight, concentration and effect of food on in vivo erosion behavior of HPMC matrix tablets. Journal of Controlled Release. 18750-58. https://doi.org/10.1016/j.jconrel.2014.04.058
2.
Fu X, Wang G, Liang W, Chow M. 2004. Prediction of drug release from HPMC matrices: effect of physicochemical properties of drug and polymer concentration. Journal of Controlled Release. 95(2):209-216. https://doi.org/10.1016/j.jconrel.2003.11.007
3.
Tassa C, Shaw SY, Weissleder R. 2011. Dextran-Coated Iron Oxide Nanoparticles: A Versatile Platform for Targeted Molecular Imaging, Molecular Diagnostics, and Therapy. Acc. Chem. Res.. 44(10):842-852. https://doi.org/10.1021/ar200084x
4.
Mittal A, Krishna, Aarti, Prasad S, Mishra PK, Sharma SK, Parshad B. Self-assembly of carbohydrate-based small amphiphiles and their applications in pathogen inhibition and drug delivery: a review. Mater. Adv.. 2(11):3459-3473. https://doi.org/10.1039/d0ma00916d
5.
Biswas R, Mondal S, Ansari MA, Sarkar T, Condiuc IP, Trifas G, Atanase LI. Chitosan and Its Derivatives as Nanocarriers for Drug Delivery. Molecules. 30(6):1297. https://doi.org/10.3390/molecules30061297
6.
Kumar L, Verma S, Joshi K, Utreja P, Sharma S. 2021. Nanofiber as a novel vehicle for transdermal delivery of therapeutic agents: challenges and opportunities. Futur J Pharm Sci. 7(1): https://doi.org/10.1186/s43094-021-00324-1
7.
Vegad U, Patel M, Khunt D, Zupančič O, Chauhan S, Paudel A. pH stimuli-responsive hydrogels from non-cellulosic biopolymers for drug delivery. Front. Bioeng. Biotechnol.. 11 https://doi.org/10.3389/fbioe.2023.1270364
8.
Tanga S, Aucamp M, Ramburrun P. Injectable Thermoresponsive Hydrogels for Cancer Therapy: Challenges and Prospects. Gels. 9(5):418. https://doi.org/10.3390/gels9050418
9.
Das S. 2021. Pectin based multi-particulate carriers for colon-specific delivery of therapeutic agents. International Journal of Pharmaceutics. 605120814. https://doi.org/10.1016/j.ijpharm.2021.120814
10.
Mazzotta E, De Benedittis S, Qualtieri A, Muzzalupo R. Actively Targeted and Redox Responsive Delivery of Anticancer Drug by Chitosan Nanoparticles. Pharmaceutics. 12(1):26. https://doi.org/10.3390/pharmaceutics12010026
11.
Federer C, Kurpiers M, Bernkop-Schnürch A. 2021. Thiolated Chitosans: A Multi-talented Class of Polymers for Various Applications. Biomacromolecules. 22(1):24-56. https://doi.org/10.1021/acs.biomac.0c00663
12.
Lutz TM, Kimna C, Casini A, Lieleg O. 2022. Bio-based and bio-inspired adhesives from animals and plants for biomedical applications. Materials Today Bio. 13100203. https://doi.org/10.1016/j.mtbio.2022.100203
13.
Mohammed M, Syeda J, Wasan K, Wasan E. An Overview of Chitosan Nanoparticles and Its Application in Non-Parenteral Drug Delivery. Pharmaceutics. 9(4):53. https://doi.org/10.3390/pharmaceutics9040053
14.
Ma J, Wang Y, Lu R. Mechanism and Application of Chitosan and Its Derivatives in Promoting Permeation in Transdermal Drug Delivery Systems: A Review. Pharmaceuticals. 15(4):459. https://doi.org/10.3390/ph15040459
15.
Gu Y, Bian Q, Zhou Y, Huang Q, Gao J. 2022. Hair follicle-targeting drug delivery strategies for the management of hair follicle-associated disorders. Asian Journal of Pharmaceutical Sciences. 17(3):333-352. https://doi.org/10.1016/j.ajps.2022.04.003
16.
Alshammari ND, Elkanayati R, Vemula SK, Al Shawakri E, Uttreja P, Almutairi M, Repka MA. Advancements in Colon-Targeted Drug Delivery: A Comprehensive Review on Recent Techniques with Emphasis on Hot-Melt Extrusion and 3D Printing Technologies. AAPS PharmSciTech. 25(7): https://doi.org/10.1208/s12249-024-02965-w
17.
Balcão VM, Vila MM. 2015. Structural and functional stabilization of protein entities: state-of-the-art. Advanced Drug Delivery Reviews. 9325-41. https://doi.org/10.1016/j.addr.2014.10.005
18.
Bashiri S, Koirala P, Toth I, Skwarczynski M. Carbohydrate Immune Adjuvants in Subunit Vaccines. Pharmaceutics. 12(10):965. https://doi.org/10.3390/pharmaceutics12100965
19.
Cao Y, Tan YF, Wong YS, Liew MWJ, Venkatraman S. Recent Advances in Chitosan-Based Carriers for Gene Delivery. Marine Drugs. 17(6):381. https://doi.org/10.3390/md17060381
20.
Masoumi Shahrbabak S, Tayebi-Khorrami V, Fadaei MR, Moezzi SMI, Hasnain MS, Nayak AK, Askari VR. 2025. Dextran-based vehicle for gene transfection: A lesson learned by nature. International Journal of Biological Macromolecules. 320145980. https://doi.org/10.1016/j.ijbiomac.2025.145980
21.
Lang S, Huang X. Carbohydrate Conjugates in Vaccine Developments. Front. Chem.. 8 https://doi.org/10.3389/fchem.2020.00284
22.
Lai J, Azad AK, Sulaiman WMAW, Kumarasamy V, Subramaniyan V, Alshehade SA. Alginate-Based Encapsulation Fabrication Technique for Drug Delivery: An Updated Review of Particle Type, Formulation Technique, Pharmaceutical Ingredient, and Targeted Delivery System. Pharmaceutics. 16(3):370. https://doi.org/10.3390/pharmaceutics16030370
23.
Salave S, Rana D, Sharma A, Bharathi K, Gupta R, Khode S, Benival D, Kommineni N. Polysaccharide Based Implantable Drug Delivery: Development Strategies, Regulatory Requirements, and Future Perspectives. Polysaccharides. 3(3):625-654. https://doi.org/10.3390/polysaccharides3030037
24.
Hasan M, Khatun A, Kogure K. Iontophoresis of Biological Macromolecular Drugs. Pharmaceutics. 14(3):525. https://doi.org/10.3390/pharmaceutics14030525
25.
Umeyor CE, Shelke V, Pol A, Kolekar P, Jadhav S, Tiwari N, Anure A, Nayak A, Bairagi G, Agale A, et al. Biomimetic microneedles: exploring the recent advances on a microfabricated system for precision delivery of drugs, peptides, and proteins. Futur J Pharm Sci. 9(1): https://doi.org/10.1186/s43094-023-00553-6
26.
Sarabia-Vallejo Á, Caja MdM, Olives AI, Martín MA, Menéndez JC. Cyclodextrin Inclusion Complexes for Improved Drug Bioavailability and Activity: Synthetic and Analytical Aspects. Pharmaceutics. 15(9):2345. https://doi.org/10.3390/pharmaceutics15092345
27.
Xu W, Ling P, Zhang T. 2013. Polymeric Micelles, a Promising Drug Delivery System to Enhance Bioavailability of Poorly Water-Soluble Drugs. Journal of Drug Delivery. 20131-15. https://doi.org/10.1155/2013/340315
28.
Ali KA, Mohin S, Mondal P, Goswami S, Ghosh S, Choudhuri S. Influence of artificial intelligence in modern pharmaceutical formulation and drug development. Futur J Pharm Sci. 10(1): https://doi.org/10.1186/s43094-024-00625-1
29.
Sarvepalli S, Pasika SR, Verma V, Thumma A, Bolla S, Nukala PK, Butreddy A, Bolla PK. A Review on the Stability Challenges of Advanced Biologic Therapeutics. Pharmaceutics. 17(5):550. https://doi.org/10.3390/pharmaceutics17050550
30.
Urru SAM, Maines E, Campomori A, Soffiati M. 2021. Safety of Sars-Cov-2 vaccines administration for adult patients with hereditary fructose intolerance. Human Vaccines & Immunotherapeutics. 17(11):4112-4114. https://doi.org/10.1080/21645515.2021.1943992
31.
Peng H, Han B, Tong T, Jin X, Peng Y, Guo M, Li B, Ding J, Kong Q, Wang Q. 2024. 3D printing processes in precise drug delivery for personalized medicine. Biofabrication. 16(3):032001. https://doi.org/10.1088/1758-5090/ad3a14