MilliporeSigma
Macrophage migration inhibitory factor is required for NLRP3 inflammasome activation.
Nature communications (2018-06-10)
Tali Lang, Jacinta P W Lee, Kirstin Elgass, Anita A Pinar, Michelle D Tate, Elizabeth H Aitken, Huapeng Fan, Sarah J Creed, Nadia S Deen, Daouda A K Traore, Ivo Mueller, Danielle Stanisic, Francesca S Baiwog, Colin Skene, Matthew C J Wilce, Ashley Mansell, Eric F Morand, James Harris
RÉSUMÉ
Macrophage migration inhibitory factor (MIF) exerts multiple effects on immune cells, as well as having functions outside the immune system. MIF can promote inflammation through the induction of other cytokines, including TNF, IL-6, and IL-1 family cytokines. Here, we show that inhibition of MIF regulates the release of IL-1α, IL-1β, and IL-18, not by affecting transcription or translation of these cytokines, but via activation of the NLRP3 inflammasome. MIF is required for the interaction between NLRP3 and the intermediate filament protein vimentin, which is critical for NLRP3 activation. Further, we demonstrate that MIF interacts with NLRP3, indicating a role for MIF in inflammasome activation independent of its role as a cytokine. These data advance our understanding of how MIF regulates inflammation and identify it as a factor critical for NLRP3 inflammasome activation.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
4-IPP, ≥97% (HPLC)