MilliporeSigma
Human Mena associates with Rac1 small GTPase in glioblastoma cell lines.
PloS one (2009-03-12)
Morihiro Higashi, Chieko Ishikawa, Jianyong Yu, Akihiro Toyoda, Hidetada Kawana, Kazuo Kurokawa, Michiyuki Matsuda, Motoo Kitagawa, Kenichi Harigaya
RÉSUMÉ
Mammarian enabled (Mena), a member of the Enabled (Ena)/Vasodilator-stimulated phosphoprotein (VASP) family of proteins, has been implicated in cell motility through regulation of the actin cytoskeleton assembly, including lamellipodial protrusion. Rac1, a member of the Rho family GTPases, also plays a pivotal role in the formation of lamellipodia. Here we report that human Mena (hMena) colocalizes with Rac1 in lamellipodia, and using an unmixing assisted acceptor depletion fluorescence resonance energy transfer (u-adFRET) analysis that hMena associates with Rac1 in vivo in the glioblastoma cell line U251MG. Depletion of hMena by siRNA causes cells to be highly spread with the formation of lamellipodia. This cellular phenotype is canceled by introduction of a dominant negative form of Rac1. A Rac activity assay and FRET analysis showed that hMena knock-down cells increased the activation of Rac1 at the lamellipodia. These results suggest that hMena possesses properties which help to regulate the formation of lamellipodia through the modulation of the activity of Rac1.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Enah