MilliporeSigma
Incidence of Staphylococcus aureus and analysis of associated bacterial communities on food industry surfaces.
Applied and environmental microbiology (2012-10-02)
Diana Gutiérrez, Susana Delgado, Daniel Vázquez-Sánchez, Beatriz Martínez, Marta López Cabo, Ana Rodríguez, Juan J Herrera, Pilar García
RÉSUMÉ
Biofilms are a common cause of food contamination with undesirable bacteria, such as pathogenic bacteria. Staphylococcus aureus is one of the major bacteria causing food-borne diseases in humans. A study designed to determine the presence of S. aureus on food contact surfaces in dairy, meat, and seafood environments and to identify coexisting microbiota has therefore been carried out. A total of 442 samples were collected, and the presence of S. aureus was confirmed in 6.1% of samples. Sixty-three S. aureus isolates were recovered and typed by random amplification of polymorphic DNA (RAPD). Profiles were clustered into four groups which were related to specific food environments. All isolates harbored some potential virulence factors such as enterotoxin production genes, biofilm formation-associated genes, antibiotic resistance, or lysogeny. PCR-denaturing gradient gel electrophoresis (PCR-DGGE) fingerprints of bacterial communities coexisting with S. aureus revealed the presence of bacteria either involved in food spoilage or of concern for food safety in all food environments. Food industry surfaces could thus be a reservoir for S. aureus forming complex communities with undesirable bacteria in multispecies biofilms. Uneven microbiological conditions were found in each food sector, which indicates the need to improve hygienic conditions in food processing facilities, particularly the removal of bacterial biofilms, to enhance the safety of food products.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 10 purifications