MilliporeSigma
  • On-line coupling of equilibrium-sorptive enrichment to gas chromatography to determine low-molecular-mass pollutants in environmental water samples.
On-line coupling of equilibrium-sorptive enrichment to gas chromatography to determine low-molecular-mass pollutants in environmental water samples.
Journal of chromatography. A (2000-02-12)
C Aguilar, H G Janssen, C A Cramers
RÉSUMÉ
On-line combination of equilibrium sorptive enrichment and gas chromatography is used for the analysis of a group of pollutants varying widely in polarity and volatility in aqueous samples at trace levels. For the ESE process open-tubular traps were used. The newly developed hyphenated method shows a high sensitivity for all the compounds under study. The detection limits were typically between 0.1 and 1 microg/l. The sample volumes required for the compounds to reach equilibrium with the stationary phase are in the range of 20 ml for the aromatic hydrocarbons included in the study (benzene, toluene and p-xylene), to 200 ml for epichlorohydrin and dichlorohydrin. Within- and between-day precision of the absolute peak areas varied between 3 and 16%. The performance of the new method was tested by the analysis of different environmental water samples.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
2,3-Dichloro-1-propanol, ≥97.0% (GC)