MilliporeSigma
  • Nonylphenols, nonylphenol-ethoxylates, linear alkylbenzenesulfonates (LAS) and bis (4-chlorophenyl)-sulfone in the German Bight of the North Sea.
Nonylphenols, nonylphenol-ethoxylates, linear alkylbenzenesulfonates (LAS) and bis (4-chlorophenyl)-sulfone in the German Bight of the North Sea.
Chemosphere (2001-11-07)
K Bester, N Theobald, H F Schröder
RÉSUMÉ
Nonylphenols and nonylphenol-ethoxylates were detected in the water and sediment samples from the German Bight of the North Sea. Additionally bis (4-chlorophenyl)-sulfone and linear alkylbenzenesulfonates (LAS) were detected in marine waters. Proof of identification is given by comparison of spectral and chromatographic data from the compounds in sample extracts to those obtained from pure standards. In extracts obtained from water samples taken in 1990 and 1995 the concentrations of nonylphenols and bis-(4-chlorophenyl)-sulfone were compared for each year. The concentrations of nonylphenols in seawater varied from 0.7 to 4.4 ng/l while in the Elbe estuary about 33 ng/l were found. In water samples taken in 1998 nonylphenol-polyethoxylates could not be determined, whereas LAS concentrations of 30 ng/l were confirmed by HPLC-MS/MS. The concentrations of bis (4-chlorophenyl)-sulfone ranged from 0.18 to 2.2 ng/l. In sediment samples LAS concentrations of 39-109 ng/g dry weight were determined.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Bis(4-chlorophenyl) sulfone, 98%