MilliporeSigma
Management of infantile subglottic hemangioma: acebutolol or propranolol?
International journal of pediatric otorhinolaryngology (2010-06-19)
Catherine Blanchet, Richard Nicollas, Michèle Bigorre, Pascal Amedro, Michel Mondain
RÉSUMÉ
The successful management of subglottic hemangioma with propranolol has been reported. We report three cases of subglottic hemangioma treated with the cardioselective beta-blocker acebutolol, 8 mg/kg/day. Treatment was efficient in two cases while an open procedure was necessary in the third child. In our experience, acebutolol could be easily administered in oral form twice-a-day only with a dose that was adaptable according to the growth of the child and showed no side effects. We also report a case of rebound growth after beta-mimetic drug use and the efficiency of propranolol treatment in such a recurrence. Considering the lack of side effects and the advantages in terms of administration, we suggest acebutolol as a first-line treatment of subglottic hemangiomas for which intervention is required.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Supelco
Acebutolol hydrochloride, analytical standard
Acebutolol hydrochloride, European Pharmacopoeia (EP) Reference Standard