MilliporeSigma
  • Trimethoprim-sulfamethoxazole-induced hyperkalemia in patients receiving inhibitors of the renin-angiotensin system: a population-based study.
Trimethoprim-sulfamethoxazole-induced hyperkalemia in patients receiving inhibitors of the renin-angiotensin system: a population-based study.
Archives of internal medicine (2010-06-30)
Tony Antoniou, Tara Gomes, David N Juurlink, Mona R Loutfy, Richard H Glazier, Muhammad M Mamdani
RÉSUMÉ
Trimethoprim therapy can cause hyperkalemia and is often coprescribed with angiotensin-converting enzyme inhibitors (ACEIs) or angiotensin receptor blockers (ARBs). The objective of this study was to characterize the risk of hyperkalemia-associated hospitalization in elderly patients who were being treated with trimethoprim-sulfamethoxazole along with either an ACEI or an ARB. We conducted a population-based, nested case-control study of a cohort of elderly patients 66 years or older who were residents of Ontario, Canada, and who were receiving continuous therapy with either an ACEI or an ARB. Case patients were those with a hyperkalemia-associated hospitalization within 14 days of receiving a prescription for trimethoprim-sulfamethoxazole, amoxicillin, ciprofloxacin, norfloxacin, or nitrofurantoin. For each case, we identified up to 4 control patients from the same cohort matched for age, sex, and presence or absence of chronic renal disease and diabetes. Odds ratios were determined for the association between hyperkalemia-associated hospitalization and previous antibiotic use. During the 14-year study period, we identified 4148 admissions involving hyperkalemia, 371 of which occurred within 14 days of antibiotic exposure. Compared with amoxicillin, the use of trimethoprim-sulfamethoxazole was associated with a nearly 7-fold increased risk of hyperkalemia-associated hospitalization (adjusted odds ratio, 6.7; 95% confidence interval, 4.5-10.0). No such risk was found with the use of comparator antibiotics. Among older patients treated with ACEIs or ARBs, the use of trimethoprim-sulfamethoxazole is associated with a major increase in the risk of hyperkalemia-associated hospitalization relative to other antibiotics. Alternate antibiotic therapy should be considered in these patients when clinically appropriate.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Supelco
Sulfaméthizol, analytical standard, ≥99% (HPLC)
Supelco
Sulfaméthizol, VETRANAL®, analytical standard