MilliporeSigma
Aggressive mammary carcinoma progression in Nrf2 knockout mice treated with 7,12-dimethylbenz[a]anthracene.
BMC cancer (2010-10-12)
Lisa Becks, Misty Prince, Hannah Burson, Christopher Christophe, Mason Broadway, Ken Itoh, Masayuki Yamamoto, Michael Mathis, Elysse Orchard, Runhua Shi, Jerry McLarty, Kevin Pruitt, Songlin Zhang, Heather E Kleiner-Hancock
RÉSUMÉ
Activation of nuclear factor erythroid 2-related factor (Nrf2), which belongs to the basic leucine zipper transcription factor family, is a strategy for cancer chemopreventive phytochemicals. It is an important regulator of genes induced by oxidative stress, such as glutathione S-transferases, heme oxygenase-1 and peroxiredoxin 1, by activating the antioxidant response element (ARE). We hypothesized that (1) the citrus coumarin auraptene may suppress premalignant mammary lesions via activation of Nrf2/ARE, and (2) that Nrf2 knockout (KO) mice would be more susceptible to mammary carcinogenesis. Premalignant lesions and mammary carcinomas were induced by medroxyprogesterone acetate and 7,12-dimethylbenz[a]anthracene treatment. The 10-week pre-malignant study was performed in which 8 groups of 10 each female wild-type (WT) and KO mice were fed either control diet or diets containing auraptene (500 ppm). A carcinogenesis study was also conducted in KO vs. WT mice (n = 30-34). Comparisons between groups were evaluated using ANOVA and Kaplan-Meier Survival statistics, and the Mann-Whitney U-test. All mice treated with carcinogen exhibited premalignant lesions but there were no differences by genotype or diet. In the KO mice, there was a dramatic increase in mammary carcinoma growth rate, size, and weight. Although there was no difference in overall survival, the KO mice had significantly lower mammary tumor-free survival. Also, in the KO mammary carcinomas, the active forms of NF-κB and β-catenin were increased ~2-fold whereas no differences in oxidized proteins were observed. Many other tumors were observed, including lymphomas. Interestingly, the incidences of lung adenomas in the KO mice were significantly higher than in the WT mice. We report, for the first time, that there was no apparent difference in the formation of premalignant lesions, but rather, the KO mice exhibited rapid, aggressive mammary carcinoma progression.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Anticorps anti-β-caténine-active (anti-ABC), clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Geranyl chloride, 95%
Sigma-Aldrich
Anti-β-Catenin Antibody, Upstate®, from rabbit
Sigma-Aldrich
Auraptene, ≥98% (HPLC)