MilliporeSigma
  • Inhibitory effects of casticin on migration of eosinophil and expression of chemokines and adhesion molecules in A549 lung epithelial cells via NF-κB inactivation.
Inhibitory effects of casticin on migration of eosinophil and expression of chemokines and adhesion molecules in A549 lung epithelial cells via NF-κB inactivation.
Journal of ethnopharmacology (2011-01-22)
Duck-jae Koh, Hong-sik Ahn, Hwan-Suck Chung, Hyojung Lee, Youngchul Kim, Jin-yong Lee, Deog-gon Kim, Moochang Hong, Minkyu Shin, Hyunsu Bae
RÉSUMÉ
The fruits of Vitex rotundifolia L. have long been used for the treatment of inflammation of the respiratory tract in East Asia. To determine if casticin, one of the constituents of Vitex rotundifolia L., has anti-allergic and anti-inflammatory effects in asthma. The in vitro anti-inflammatory activity of casticin was studied in A549 human type II-like epithelial lung cells using an eotaxin inhibition assay. Additionally, its effects on eotaxin, regulated on activation normal T cell expressed and secreted (RANTES), vascular cell adhesion molecule (VCAM)-1, and inter-cellular adhesion molecule (ICAM)-1 expression were investigated by real time-polymerase chain reaction (real time-PCR). The inhibition of nuclear factor κB (NF-κB) activity in the presence of casticin was determined by analyzing confocal microscopy images of fluorescence immunocytochemical analysis while the suppression of inhibitory κB (IκB)-α phosphorylation was studied using Western blot analysis. Finally, the inhibitory effect of casticin on eosinophil migration toward prestimulated A549 cell media was measured using the human eosinophilic leukemia cell line. Casticin significantly suppressed eotaxin production in cytokine activated A549 lung epithelial cells. Casticin also suppressed the mRNA expression levels of eotaxin, RANTES, VCAM-1, and ICAM-1, which subsequently contributed to the inhibition of eosinophil migration. Furthermore, casticin inhibited IκB-α phosphorylation and nuclear translocation of p65 in A549 cells. Casiticin inhibited the eosinophil migration and activity of chemokines and adhesion molecules involved in the inflammatory process of asthma by suppressing the NF-κB pathway. These results suggest that casticin has the potential for use in the treatment of allergic asthma.