MilliporeSigma
[Allergic contact dermatitis to synthetic rubber, neoprene in compression stockings].
Masui. The Japanese journal of anesthesiology (2011-02-26)
Ju Mizuno, Hiroshi In-Nami
RÉSUMÉ
Compression stockings are used for patients under general anesthesia to prevent occurrence of deep venous thrombosis. We report a case of allergic contact dermatitis to synthetic rubber, neoprene in compression stockings. A 53-year-old house wife had a history of sensitivity like skin eruption and disstasis to rubber products such as rubber band. Left nephrectomy for rupture of renal angiomyolipoma was scheduled under general and epidural anesthesia. Further examination for gum allergy was not performed before the operation, although latex allergy was suspected. The operation was performed uneventfully under latex-safe environment in the operating room under guideline for latex allergy. Postoperatively, ringed edematous erythema and wheal occurred in her bilateral thighs compressed with the upper part of compression stockings. The skin symptoms continued for more than four days. After disappearance of the skin symptoms, she was discharged from the hospital on the ninth day after the operation. Synthetic rubber, neoprene, in the upper part of compression stockings to prevent slipping down might cause allergic contact dermatitis. We should take care of occurrence of allergic contact dermatitis to synthetic rubber, neoprene in compression stockings in patients with rubber allergy.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Polychloroprene, 85% trans, 10% cis