MilliporeSigma
Synthesis and evaluation of analogues of HYNIC as bifunctional chelators for technetium.
Dalton transactions (Cambridge, England : 2003) (2011-02-26)
Levente K Meszaros, Anica Dose, Stefano C G Biagini, Philip J Blower
RÉSUMÉ
6-Hydrazinonicotinic acid (HYNIC, 1) is a well-established bifunctional technetium-binding ligand often used to synthesise bioconjugates for radiolabelling with Tc-99m. It is capable of efficient capture of technetium at extremely low concentrations, but the structure of the labelled complexes is heterogeneous and incompletely understood. In particular, it is of interest to determine whether, at the no-carrier-added level, it acts in a chelating or non-chelating mode. Here we report two new isomers of HYNIC: 2-hydrazinonicotinic acid (2-HYNIC, 2), which (like 1) is capable of chelation through the mutually ortho hydrazine and pyridine nitrogens and 4-hydrazinonicotinic acid (4-HYNIC, 3), which is not (due to the para-relationship of the hydrazine and pyridine nitrogens). LC-MS shows that the coordination chemistry of 2 with technetium closely parallels that of conventional 1, and no advantages of one over the other in terms of potential labelling efficiency or isomerism were discernable. Both 1 and 2 formed complexes with the loss of 5 protons from the ligand set, whether the co-ligand was tricine or EDDA. Ligand 3, however, failed to complex technetium except at very high ligand concentration: the marked contrast with 1 and 2 suggests that chelation, rather than nonchelating coordination, is a key feature of technetium coordination by HYNIC. Two further new HYNIC analogues, 2-chloro-6-hydrazinonicotinic acid (2-chloro-HYNIC, 4a) and 2,6-dihydrazinonicotinic acid (diHYNIC, 5) were also synthesised. The coordination chemistry of 4a with technetium was broadly parallel to that of 1 and 2 although it was a less efficient chelator, while 5 also behaved as an efficient chelator of technetium, but its coordination chemistry remains poorly defined and requires further investigation before it can sensibly be adopted for (99m)Tc-labelling. The new analogues 4a and 5 present an opportunity to develop trifunctional HYNIC analogues for more complex bioconjugate synthesis.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Ethylenediamine-N,N′-diacetic acid, ≥98%