MilliporeSigma
  • Radical-mediated nitrile translocation as the key step in the stereoselective transformation of 2-(4-chloro-2-cyanobutyl)aziridines to methyl cis-(1-arylmethyl-4-phenylpiperidin-2-yl)acetates.
Radical-mediated nitrile translocation as the key step in the stereoselective transformation of 2-(4-chloro-2-cyanobutyl)aziridines to methyl cis-(1-arylmethyl-4-phenylpiperidin-2-yl)acetates.
Organic & biomolecular chemistry (2012-03-14)
Karel Vervisch, Matthias D'hooghe, Karl W Törnroos, Norbert De Kimpe
RÉSUMÉ
Non-activated 2-(4-chloro-2-cyano-2-phenylbutyl)aziridines were used as building blocks for the stereoselective synthesis of novel cis-2-cyanomethyl-4-phenylpiperidines via a microwave-assisted aziridine to piperidine ring expansion followed by a radical-induced nitrile translocation through initial formation and subsequent cleavage of intermediate bicyclic iminyl radicals. Furthermore, these 2-(cyanomethyl)piperidines were shown to be eligible substrates for the preparation of methyl cis-(1-arylmethyl-4-piperidin-2-yl)acetates through a Pinner reaction using gaseous HCl in methanol.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
4-Phenylpiperidine, 97%