MilliporeSigma
American trypanosomiasis (Chagas disease).
Infectious disease clinics of North America (2012-05-29)
Anis Rassi, Anis Rassi, Joffre Marcondes de Rezende
RÉSUMÉ
Chagas disease, also known as American trypanosomiasis, is a chronic infection caused by Trypanosoma cruzi, a protozoan parasite. It is transmitted to human beings mainly through the feces of infected triatomine bugs. The disease affects an estimated 8 to 10 million people in the Americas, putting them at risk of developing life-threatening cardiac and gastrointestinal complications. This article provides a brief update on the epidemiology, clinical manifestations, diagnosis, and treatment of Chagas disease.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Nifurtimox, ≥98% (HPLC)