MilliporeSigma
  • Photophysical and photochemical properties of a novel thiol terminated low symmetry zinc phthalocyanine complex and its gold nanoparticles conjugate.
Photophysical and photochemical properties of a novel thiol terminated low symmetry zinc phthalocyanine complex and its gold nanoparticles conjugate.
Dalton transactions (Cambridge, England : 2003) (2013-02-07)
Thandekile P Mthethwa, Sinem Tuncel, Mahmut Durmuş, Tebello Nyokong
RÉSUMÉ
A novel thiol functionalized zinc phthalocyanine complex (ZnPcSH) is reported in this work. This complex was conjugated to gold nanoparticles. The photophysical and photochemical properties of the complex and the conjugate were investigated. Upon conjugation a blue shift was observed from the UV-Vis spectra. The conjugate showed a decrease in the fluorescence quantum yield and lifetime. An increase in the triplet quantum yield and lifetime was observed for ZnPcSH following conjugation to gold nanoparticles. Both ZnPcSH and its conjugate with gold nanoparticles showed high singlet oxygen quantum yields with the conjugate being higher than the Pc alone.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Zinc phthalocyanine, Dye content 97 %