MilliporeSigma
[The sensitivity to antibiotics and chloramine B of opportunistic microorganisms isolated from patients].
Mikrobiologicheskii zhurnal (1989-11-01)
N I Balakliets, T I Balakliets, A Ia Tsyganenko, F Ia Partina, V V Zozulia
RÉSUMÉ
Strains of staphylococci, klebsiella, proteins, blue pus bacillus and enterobacteria resistant not only to antibiotics but also to chloramine, a disinfectant, were isolated from the test material samples obtained from patients with different purulent-inflammatory processes. The following strains are revealed among the test microorganisms: sensitive simultaneously to antibiotics and chloramine; sensitive to antibiotics but resistant to chloramine; sensitive to chloramine but resistant to antibiotics; simultaneously resistant both to antibiotics and chloramine.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
N-Chlorobenzenesulfonamide sodium salt, ~28% active chlorine basis