MilliporeSigma
alpha, beta-Bidentate CrADP abolishes the negative cooperativity of yeast mitochondrial F1-ATPase.
Biochimica et biophysica acta (1985-01-23)
H J Wieker, B Hess
RÉSUMÉ
The hydrolysis of MgATP and MgITP by mitochondrial F1-ATPase from Saccharomyces cerevisiae is competitively inhibited by alpha, beta-CrADP, alpha, beta, gamma-CrATP and beta, gamma-CrATP. The apparent K1 values of the three complexes are in the range of the half-saturating MgATP concentration. The negative cooperativity (nH = 0.7) of MgATP hydrolysis is totally abolished by alpha, beta-CrADP (nH = 1.0), while it is not affected by the CrATP. It is concluded that alpha, beta-CrADP binds exclusively at the regulatory site and that CrATP binds exclusively to the catalytic site.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Inosine 5′-triphosphate trisodium salt, ≥95%