MilliporeSigma
The use of D-penicillamine in patients with rheumatoid arthritis undergoing hemodialysis.
Clinical nephrology (1986-05-01)
F Matthey, D Perrett, R N Greenwood, L R Baker
RÉSUMÉ
D-penicillamine and its major metabolites cysteine-penicillamine disulphide (CP) and penicillamine disulphide (P2) concentrations were measured in plasma from a hemodialysis patient with rheumatoid arthritis. CP and P2 alone were measured in plasma and a plasma ultrafiltrate from a second patient. On penicillamine 250 mg thrice weekly taken after dialysis pre-dialysis penicillamine concentrations were in the range 5.9-9.9 mumol/l. CP and P2 concentrations remained stable (range 139-197 mumol/l and 10-20 mumol/l) over 5 weeks and were of the same order as previously found in patients with normal renal function on higher doses of the drug. On penicillamine 250 mg daily concentrations of metabolites CP and P2 reach 193 mumol/l and 59.2 mumol/l after 2 and 3 weeks respectively. Concentration of metabolites fell by about half and of penicillamine by about a third after dialysis. Concentration of metabolites in ultrafiltrate were on average 75% lower than in plasma. Penicillamine 250 mg thrice weekly given after dialysis appears to be an appropriate dose for hemodialysis patients with rheumatoid arthritis.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
D-Penicillamine disulfide, 97%