MilliporeSigma
  • [Antiviral action of the macromolecular derivative of the platinum with deoxyribonucleic acid in the treatment of patient with malignant liver neoplasm].
[Antiviral action of the macromolecular derivative of the platinum with deoxyribonucleic acid in the treatment of patient with malignant liver neoplasm].
Klinicheskaia khirurgiia (1994-01-01)
S A Shalimov, L V Keĭsevich, E B Medvetskiĭ, B A Tarasiuk, I I Volchenskova
RÉSUMÉ
Results of the action of a preparation consisting of cis-dichlorodiammineplatinum and deoxyribonucleic acid combined in the saline balanced solution by means of the original technique on cytomegalovirus infection of patients with cancer of the liver are presented. Clearly pronounced antivirus activity of the preparation developed is noted.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Dichloro(ethylenediamine)platinum(II), 99%