MilliporeSigma
Chemical dehydration for rapid paraffin embedding.
Biotechnic & histochemistry : official publication of the Biological Stain Commission (1994-09-01)
W Möller, G Möller
RÉSUMÉ
We describe chemical dehydration with 2,2-dimethoxypropane (DMP) for rapid paraffin embedding using a mixture of DMP and mineral oil followed by mineral oil as clearing intermediates. This method is useful for classical histological techniques as well as for histochemistry and immunocytochemistry.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
2,2-Dimethoxypropane, reagent grade, 98%
Supelco
2,2-Dimethoxypropane, analytical standard