MilliporeSigma
  • Tau protein is phosphorylated by cyclic AMP-dependent protein kinase and calcium/calmodulin-dependent protein kinase II within its microtubule-binding domains at Ser-262 and Ser-356.
Tau protein is phosphorylated by cyclic AMP-dependent protein kinase and calcium/calmodulin-dependent protein kinase II within its microtubule-binding domains at Ser-262 and Ser-356.
The Biochemical journal (1996-06-01)
J M Litersky, G V Johnson, R Jakes, M Goedert, M Lee, P Seubert
RÉSUMÉ
Phosphorylation of tau protein at Ser-262 has been shown to diminish its ability to bind to taxol-stabilized microtubules. The paired helical filaments (PHFs) found in Alzheimer's disease brain are composed of PHF-tau, which is hyperphosphorylated at multiple sites including Ser-262. However, protein kinase(s) able to phosphorylate this site are still under investigation. In this study, the ability of cyclic AMP-dependent protein kinase (cAMP-PK) and calcium/calmodulin-dependent protein kinase II (CaMKII) to phosphorylate tau at Ser-262, as well as Ser-356, is demonstrated by use of a monoclonal antibody (12E8) which has been shown to recognize tau when these sites are phosphorylated. Cleavage of cAMP-PK-phosphorylated tau at cysteine residues by 2-nitro-5-thiocyanobenzoic acid, which cuts the protein into essentially two fragments and separates Ser-262 from Ser-356, revealed that cAMP-PK phosphorylates both Ser-262 and Ser-356. In addition, phosphorylation with cAMP-PK or CaMKII of recombinant tau in which Ser-262, Ser-356 or both had been mutated to alanines, clearly demonstrated that cAMP-PK and CaMKII were able to phosphorylate both sites. Mitogen-activated protein kinase or protein kinase C did not phosphorylate tau at Ser-262 and/or Ser-356. Finally, evidence is presented that phosphorylation of both these sites occurs in cultured nerve cells under certain conditions, indicating their potential physiological relevance.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
2-Nitro-5-thiocyanatobenzoic acid, powder