MilliporeSigma
Use of 5-nitroindole-2'-deoxyribose-5'-triphosphate for labelling and detection of oligonucleotides.
Nucleosides & nucleotides (1998-08-26)
C L Smith, A C Simmonds, A L Hamilton, D L Martin, A G Lashford, D Loakes, F Hill, D M Brown
RÉSUMÉ
The 5'-triphosphate of 5-nitroindole-2'-deoxyriboside has been shown to be a good substrate for terminal deoxynucleotidyl transferase (TdT). An antibody has been prepared for the detection of 5-nitroindole and has been used for the detection of 5-nitroindole tailed DNA both in single-stranded form and after hybridisation to a template. This is therefore a new method for the detection of nucleic acid probes.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
5-Nitroindole, 98%