MilliporeSigma
Novel cytokine release inhibitors. Part IV: analogs of podocarpic acid.
Bioorganic & medicinal chemistry letters (1999-03-26)
W He, A Gavai, F C Huang, J Regan, B Hanney, G Poli, J Bruno, W K Chan, S W Djuric, K T Yu, A Zilberstein
RÉSUMÉ
Podocarpic acid derivatives as cytokine (IL-1beta) release inhibitors are discussed.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Podocarpic acid, 98%