MilliporeSigma
Gramicidin toxicity in NG108-15 cells: protective effects of acetamidine and guanidine.
Cell biology and toxicology (2000-05-17)
J A Doebler
RÉSUMÉ
Studies were conducted using a novel in vitro approach to investigate the efficacy of acetamidine hydrochloride (ACE) and guanidine hydrochloride (GUAN), previously shown to block gramicidin D (GRAM) channels in artificial membranes, in preventing the toxic effects of GRAM in NG108-15 (neuroblastoma x glioma hybrid) cells. Specifically, intracellular microelectrode techniques were employed to examine changes in membrane resting potential (Vm) and input resistance (Rin). At 1 micromol/L, ACE significantly reduced loss of Vm induced by 1 or 10 microg/ml GRAM, although higher concentrations of ACE did not afford enhanced antagonism. GUAN, in contrast, produced a concentration-dependent antagonism of GRAM-induced Vm and Rin loss, with high concentrations (10 or 100 micromol/L) completely preventing diminutions in both Vm and Rin. In control cells superfused without GRAM, ACE produced a direct, concentration-dependent reduction in Vm and Rin, whereas GUAN hyperpolarized NG108-15 cells but did not alter Rin. These data represent the initial demonstration of the reversal of GRAM toxicity in an intact cell system.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Acetamidine hydrochloride, 95%