MilliporeSigma
  • Beta2- and beta3-adrenoceptors activate glucose uptake in chick astrocytes by distinct mechanisms: a mechanism for memory enhancement?
Beta2- and beta3-adrenoceptors activate glucose uptake in chick astrocytes by distinct mechanisms: a mechanism for memory enhancement?
Journal of neurochemistry (2007-08-08)
Dana S Hutchinson, Roger J Summers, Marie E Gibbs
RÉSUMÉ
Isoprenaline, acting at beta-adrenoceptors (ARs), enhances memory formation in single trial discriminated avoidance learning in day-old chicks by mechanisms involving alterations in glucose and glycogen metabolism. Earlier studies of memory consolidation in chicks indicated that beta3-ARs enhanced memory by increasing glucose uptake, whereas beta2-ARs enhance memory by increasing glycogenolysis. This study examines the ability of beta-ARs to increase glucose uptake in chick forebrain astrocytes. The beta-AR agonist isoprenaline increased glucose uptake in a concentration-dependent manner, as did insulin. Glucose uptake was increased by the beta2-AR agonist zinterol and the beta3-AR agonist CL316243, but not by the beta1-AR agonist RO363. In chick astrocytes, reverse transcription-polymerase chain reaction studies showed that beta1-, beta2-, and beta3-AR mRNA were present, whereas radioligand-binding studies showed the presence of only beta2- and beta3-ARs. beta-AR or insulin-mediated glucose uptake was inhibited by phosphatidylinositol-3 kinase and protein kinase C inhibitors, suggesting a possible interaction between the beta-AR and insulin pathways. However beta2- and beta3-ARs increase glucose uptake by two different mechanisms: beta2-ARs via a Gs-cAMP-protein kinase A-dependent pathway, while beta3-ARs via interactions with Gi. These results indicate that activation of beta2- and beta3-ARs causes glucose uptake in chick astrocytes by distinct mechanisms, which may be relevant for memory enhancement.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Zinterol hydrochloride, ≥98% (HPLC)