MilliporeSigma
Manipulation of lipid bioaccessibility of almond seeds influences postprandial lipemia in healthy human subjects.
The American journal of clinical nutrition (2008-10-10)
Sarah E E Berry, Elizabeth A Tydeman, Hannah B Lewis, Ravneet Phalora, Jennifer Rosborough, David R Picout, Peter R Ellis
RÉSUMÉ
Plant cell walls are known to influence the rate and extent of lipid release from plant food tissues during digestion; however, the effect of cell wall structure on postprandial lipemia is unknown. The objective was to investigate the effects of lipid release (bioaccessibility) on postprandial lipemia by comparing lipid encapsulated by cell walls with lipid present as free oil. A randomized crossover trial (n = 20 men) compared the effects of 3 meals containing 54 g fat provided as whole almond seed macroparticles (WA), almond oil and defatted almond flour (AO), or a sunflower oil blend as control (CO) on postprandial changes in oxidative stress (8-isoprostane F(2)alpha concentrations), vascular tone (peripheral augmentation index), and plasma triacylglycerol, glucose, and insulin concentrations. The postprandial increase in plasma triacylglycerol was lower [74% and 58% lower incremental area under curve (iAUC)] after the WA meal than after the AO and CO meals (P < 0.001). Increases in plasma glucose concentrations (0-180 min) were significantly higher after the WA meal (iAUC: 114; 95% CI: 76, 153) than after the AO meal (iAUC: 74; 95% CI: 48, 99) (P < 0.05), but no significant differences from the CO meal were observed (iAUC: 88; 95% CI: 66, 109). The peak reductions in peripheral augmentation index after the WA, AO, and CO meals (-9.5%, -10.1%, and -12.6%, respectively, at 2 h) were not significantly different between meals. Plasma 8-isoprostane F(2)alpha and insulin concentrations did not differ significantly between meals. The bioaccessibility of lipid in almond seeds, which is regulated by the structure and properties of cell walls, plays a primary role in determining postprandial lipemia.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Almond oil from Prunus dulcis, tested according to Ph. Eur.