MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Inter-laboratory study of an LC-MS/MS method for simultaneous determination of deoxynivalenol and its acetylated derivatives, 3-acetyl-deoxynivalenol and 15-acetyl-deoxynivalenol in wheat.
Inter-laboratory study of an LC-MS/MS method for simultaneous determination of deoxynivalenol and its acetylated derivatives, 3-acetyl-deoxynivalenol and 15-acetyl-deoxynivalenol in wheat.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2013-03-09)
Tomoya Yoshinari, Toshitsugu Tanaka, Eiichi Ishikuro, Masakazu Horie, Toshihiro Nagayama, Masahiro Nakajima, Shigehiro Naito, Takahiro Ohnishi, Yoshiko Sugita-Konishi
RÉSUMÉ
To validate an LC-MS/MS method for simultaneous determination of deoxynivalenol (DON) and its acetylated derivatives, 3-acetyl-deoxynivalenol (3ADON) and 15-acetyl-deoxynivalenol (15ADON), in wheat using a multifunctional column, an inter-laboratory study was performed in 9 laboratories using one blank wheat sample, three spiked wheat samples (10, 50, 150 µg/kg) and one naturally contaminated wheat sample. The recoveries ranged from 98.8 to 102.6% for DON, 89.3 to 98.7% for 3ADON, and from 84.9 to 90.0% for 15ADON. The relative standard deviations for repeatability (RSDR) and reproducibility (RSDR) of DON were in the ranges of 7.2-11.3% and 9.5-22.6%, respectively. For 3ADON, the RSDR ranged from 5.3 to 9.5% and the RSDR ranged from 16.1 to 18.0%, while for 15ADON, the RSDR ranged from 6.2 to 11.2% and the RSDR ranged from 17.0 to 27.2%. The HorRat values for the three analytes ranged from 0.4 to 1.2. These results validate this method for the simultaneous determination of DON and its acetylated derivatives, 3ADON and 15ADON.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Désoxynivalénol
Supelco
Deoxynivalenol solution, 100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
15-O-Acetyl-4-deoxynivalenol from Fusarium graminearum
Sigma-Aldrich
3-Acetyldeoxynivalenol from Fusarium roseum, from Fusarium roseum
4-Deoxynivalenol in acetonitrile, IRMM®, certified reference material
Supelco
3-Acetyldeoxynivalenol solution, ~100 μg/mL in acetonitrile, analytical standard
Supelco
15-Acetyldeoxynivalenol solution, ~100 μg/mL in acetonitrile, analytical standard