MilliporeSigma
Gas-chromatographic analysis of busulfan for therapeutic drug monitoring.
Cancer chemotherapy and pharmacology (1993-01-01)
L Embree, R B Burns, J R Heggie, G L Phillips, D E Reece, J J Spinelli, D O Hartley, N J Hudon, J H Goldie
RÉSUMÉ
The development and validation of a gas chromatographic assay method for determination of total and free busulfan concentrations in human plasma for pharmacokinetic studies is reported. 1,6-Bis(methanesulfonyloxy)hexane, the internal standard, and a potential metabolite, 3-hydroxysulfolane, were synthesized. Plasma and plasma ultrafiltrate samples containing busulfan and internal standard were extracted with ethyl acetate and derivatized with 2,3,5,6-tetrafluorothiophenol prior to gas chromatographic determination. The 63Ni electron-capture detector provided a limit of detection of 0.0600 microgram/ml with a limit of quantitation of 0.100 microgram/ml busulfan in biological samples. Calibration curves were linear from 0.100 to 3.00 micrograms/ml in plasma (500 microliters) and 0.100 to 2.00 micrograms/ml in plasma ultrafiltrate (100 microliters). Extraction and derivatization yields ranged from 78.4% to 89.6% and 56.0% to 71.3%, respectively. Specificity of this assay for busulfan in the presence of its potential metabolites was demonstrated. Also, plasma samples containing co-administered drugs gave no response under these conditions. Clinical samples obtained following administration of a 1 mg/kg oral busulfan dose demonstrate the applicability of this method to analysis of total and free plasma concentrations.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
2,3,5,6-Tetrafluorobenzenethiol, 97%