MilliporeSigma
In vitro enhancement of the mutagenicity of 4-nitro-o-phenylenediamine by plant S-9.
Environmental mutagenesis (1985-01-01)
J M Gentile, G J Gentile, S Townsend, M J Plewa
RÉSUMÉ
The direct-acting mutagen 4-nitro-o-phenylenediamine (NOP) was activated in vitro by pea or tobacco S-9 into a more potent mutagen in Salmonella typhimurium strain TA98. NOP mutagenicity was not altered by Aroclor 1254-induced rat liver S-9. The plant S-9 activation of NOP was heat-sensitive but was not NADPH-dependent, did not involve superoxide radicals, and was not inhibited by CO. A direct relationship between plant peroxidase and NOP activation was established. Several purified peroxidases including horseradish peroxidase, chloroperoxidase, and lactoperoxidase also activated NOP. The perodoxidative process was not H2O2-dependent but was partially inhibited by a peroxidase inhibitor.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
4-Nitro-o-phenylenediamine, 98%