MilliporeSigma
Synthesis and Diels-Alder reactivity of ortho-carbazolequinones.
Chemical & pharmaceutical bulletin (2004-09-02)
Muriel Compain-Batissou, Djamila Latreche, Jacques Gentili, Nadia Walchshofer, Zouhair Bouaziz
RÉSUMÉ
Oxidation of 2- and 3-hydroxycarbazoles with Frémy's salt gave the corresponding ortho-carbazolequinones. These molecules react as carbodienophiles in Diels-Alder reaction with 1-acetoxy-1,3-butadiene and 1,3-cyclopentadiene to provide the novel benzocarbazolequinone structures 15, 16, 18 and 19.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
1-Acetoxy-1,3-butadiene, mixture of cis and trans