MilliporeSigma
Ephs and ephrins in cancer: ephrin-A1 signalling.
Seminars in cell & developmental biology (2011-11-02)
Amanda Beauchamp, Waldemar Debinski
RÉSUMÉ
Ephrin-A1 and its primary receptor, EphA2, are involved in numerous physiological processes and have been intensely studied for their roles in malignancy. Ephrin-Eph signalling is complex on its own and is also cell-type dependent, making elucidation of the exact role of ephrin-A1 in neoplasia challenging. Multiple oncogenic signalling pathways, such as MAP/ERK and PI3K are affected by ephrin-A1, and in some cases evidence suggests the promotion of a specific pathway in one cell or cancer type and inhibition of the same pathway in another type of cell or cancer. Ephrin-A1 also plays an integral role in angiogenesis and tumor neovascularization. Until recently, studies investigating ephrins focused on the ligands as GPI-anchored proteins that required membrane anchoring or artificial clustering for Eph receptor activation. However, recent studies have demonstrated a functional role for soluble, monomeric ephrin-A1. This review will focus on various forms of ephrin-A1-specific signalling in human malignancy.