MilliporeSigma
Gene expression profiling of murine T-cell lymphoblastic lymphoma identifies deregulation of S-phase initiating genes.
Leukemia research (2013-07-31)
Magdalena Julia Dabrowska, Ditte Ejegod, Louise Berkhoudt Lassen, Hans Erik Johnsen, Matthias Wabl, Finn Skou Pedersen, Karen Dybkær
RÉSUMÉ
In a search for genes and pathways implicated in T-cell lymphoblastic lymphoma (T-LBL) development, we used a murine lymphoma model, where mice of the NMRI-inbred strain were inoculated with murine leukemia virus mutants. The resulting tumors were analyzed by integration analysis and global gene expression profiling to determine the effect of the retroviral integrations on the nearby genes, and the deregulated pathways in the tumors. Gene expression profiling identified increased expression of genes involved in the minichromosome maintenance and origin of recognition pathway as well as downregulation in negative regulators of G1/S transition, indicating increased S-phase initiation in murine T-LBLs.