MilliporeSigma
Tumor growth delay by adjuvant alternating electric fields which appears non-thermally mediated.
Bioelectrochemistry (Amsterdam, Netherlands) (2015-05-09)
Quim Castellví, Mireia M Ginestà, Gabriel Capellà, Antoni Ivorra
RÉSUMÉ
Delivery of the so-called Tumor Treatment Fields (TTFields) has been proposed as a cancer therapy. These are low magnitude alternating electric fields at frequencies from 100 to 300 kHz which are applied continuously in a non-invasive manner. Electric field delivery may produce an increase in temperature which cannot be neglected. We hypothesized that the reported results obtained by applying TTFields in vivo could be due to heat rather than to electrical forces as previously suggested. Here, an in vivo study is presented in which pancreatic tumors subcutaneously implanted in nude mice were treated for a week either with mild hyperthermia (41 °C) or with TTFields (6 V/cm, 150 kHz) and tumor growth was assessed. Although the TTFields applied singly did not produce any significant effect, the combination with chemotherapy did show a delay in tumor growth in comparison to animals treated only with chemotherapy (median relative reduction=47%). We conclude that concomitant chemotherapy and TTFields delivery show a beneficial impact on pancreatic tumor growth. Contrary to our hypothesis, this impact is non-related with the induced temperature increase.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human MKI67