MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Antimicrobial peptides (AMPs) produced by Saccharomyces cerevisiae induce alterations in the intracellular pH, membrane permeability and culturability of Hanseniaspora guilliermondii cells.
Antimicrobial peptides (AMPs) produced by Saccharomyces cerevisiae induce alterations in the intracellular pH, membrane permeability and culturability of Hanseniaspora guilliermondii cells.
International journal of food microbiology (2015-04-22)
Patrícia Branco, Tiago Viana, Helena Albergaria, Nils Arneborg
RÉSUMÉ
Saccharomyces cerevisiae produces antimicrobial peptides (AMPs) during alcoholic fermentation that are active against several wine-related yeasts (e.g. Hanseniaspora guilliermondii) and bacteria (e.g. Oenococcus oeni). In the present study, the physiological changes induced by those AMPs on sensitive H. guilliermondii cells were evaluated in terms of intracellular pH (pHi), membrane permeability and culturability. Membrane permeability was evaluated by staining cells with propidium iodide (PI), pHi was determined by a fluorescence ratio imaging microscopy (FRIM) technique and culturability by a classical plating method. Results showed that the average pHi of H. guilliermondii cells dropped from 6.5 (healthy cells) to 5.4 (damaged cells) after 20 min of exposure to inhibitory concentrations of AMPs, and after 24 h 77.0% of the cells completely lost their pH gradient (∆pH=pHi-pHext). After 24h of exposure to AMPs, PI-stained (dead) cells increased from 0% to 77.7% and the number of viable cells fell from 1×10(5) to 10 CFU/ml. This means that virtually all cells (99.99%) became unculturable but that a sub-population of 22.3% of the cells remained viable (as determined by PI staining). Besides, pHi results showed that after 24h, 23% of the AMP-treated cells were sub-lethally injured (with 0<∆pH<3). Taken together, these results indicated that this subpopulation was under a viable but non-culturable (VBNC) state, which was further confirmed by recuperation assays. In summary, our study reveals that these AMPs compromise the plasma membrane integrity (and possibly also the vacuole membrane) of H. guilliermondii cells, disturbing the pHi homeostasis and inducing a loss of culturability.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Diméthylsulfoxyde, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Diméthylsulfoxyde, Molecular Biology
Sigma-Aldrich
Diméthylsulfoxyde, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Diméthylsulfoxyde, anhydrous, ≥99.9%
Sigma-Aldrich
Diméthylsulfoxyde, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Iodure de propidium, ≥94.0% (HPLC)
Sigma-Aldrich
Acide citrique, ACS reagent, ≥99.5%
Sigma-Aldrich
Diméthylsulfoxyde, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Acide citrique, ≥99.5%, FCC, FG
Sigma-Aldrich
Acide citrique, 99%
Sigma-Aldrich
Acide citrique, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Diméthylsulfoxyde, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Iodure de propidium solution
Sigma-Aldrich
Diméthylsulfoxyde, PCR Reagent
Sigma-Aldrich
Acide citrique, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Acide citrique, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Diméthylsulfoxyde, Vetec, reagent grade, 99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Diméthylsulfoxyde, ≥99.6%, ReagentPlus®