MilliporeSigma
The GSTA1 polymorphism and cyclophosphamide therapy outcomes in lupus nephritis patients.
Clinical immunology (Orlando, Fla.) (2015-07-30)
Hong-Na Wang, Xiao-Ye Zhu, Ying Zhu, Qiong-Hong Xie, Lin-Yun Lai, Miao Zhao, Yuan-Cheng Chen, Jun Xue, Chuan-Ming Hao, Yong Gu, Shan-Yan Lin
RÉSUMÉ
Pulsed low-dose cyclophosphamide (CTX) therapy has become a very effective approach in improving the clinical outcomes of lupus nephritis (LN) patients. However, variations of CTX therapeutic outcomes in LN patients are incompletely understood. We investigated the contributions of known allelic variants to CTX therapy outcomes in 77 LN patients. Then, 22 out of the 77 patients were randomly enrolled to evaluate the pharmacokinetic profiles. LN patients with a GSTA1*A mutation (CT heterozygous) had more risk of non-remission (44% vs. 20%, P=0.005). Pharmacokinetic data indicated that patients with a GSTA1*A heterozygous variant had a lower exposure to 4-hydroxycyclophosphamide (4OHCTX) compared to wild-type patients (AUC4OHCTX: 12.8 (9.8, 19.5) vs. 27.5 (18.1, 32.8) h mg/l, P=0.023). Clinical remission was significantly related to higher exposure of 4OHCTX (P=0.038). In conclusion, LN patients with GSTA1*A heterozygous genotypes had poor CTX treatment remission due to less exposure to activated metabolites of CTX.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
GST A1-1, Recombinant Human