MilliporeSigma
Beta-synuclein regulates Akt activity in neuronal cells. A possible mechanism for neuroprotection in Parkinson's disease.
The Journal of biological chemistry (2004-03-18)
Makoto Hashimoto, Pazit Bar-On, Gilbert Ho, Takato Takenouchi, Edward Rockenstein, Leslie Crews, Eliezer Masliah
RÉSUMÉ
Recent studies have shown that the neurodegenerative process in disorders with Lewy body formation, such as Parkinson's disease and dementia with Lewy bodies, is associated with alpha-synuclein accumulation and that beta-synuclein might protect the central nervous system from the neurotoxic effects of alpha-synuclein. However, the mechanisms are unclear. The main objective of the present study was to investigate the potential involvement of the serine threonine kinase Akt (also known as protein kinase B) signaling pathway in the mechanisms of beta-synuclein neuroprotection. For this purpose, Akt activity and cell survival were analyzed in synuclein-transfected B103 neuroblastoma cells and primary cortical neurons. Beta-synuclein transfection resulted in increased Akt activity and conferred protection from the neurotoxic effects of rotenone. Down-regulation of Akt expression resulted in an increased susceptibility to rotenone toxicity, whereas transfection with a lentiviral vector encoding for beta-synuclein was protective. The effects of beta-synuclein on the Akt pathway appear to be by direct interaction between these molecules and were independent of upstream signaling molecules. Taken together, these results indicate that the mechanisms of beta-synuclein neuroprotection might involve direct interactions between beta-synuclein and Akt and suggest that this signaling pathway could be a potential therapeutic target for neurological conditions associated with parkinsonism and alpha-synuclein aggregation.