MilliporeSigma
Gas-phase interactions of organotin compounds with cysteine.
Journal of mass spectrometry : JMS (2016-11-04)
Latifa Latrous, Jean-Yves Salpin, Violette Haldys, Emmanuelle Léon, Catarina Correia, Al Mokhtar Lamsabhi
RÉSUMÉ
The gas-phase interactions of cysteine with di-organotin and tri-organotin compounds have been studied by mass spectrometry experiments and quantum calculations. Positive-ion electrospray spectra show that the interaction of di- and tri-organotins with cysteine results in the formation of [(R)
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Diphenyltin dichloride, 96%